Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
This item is not returnable.
View return policy
Description
- For medium- to high-volume critical care testing sites
- Full test menu from a small blood sample
- Self-contained, easily replaceable reagent and wash cartridges
- Easy-to-use system features three simple steps
- Enhanced system security with McAfee Embedded Control, firewall and verified system connection
- Immediate results available in just 1 minute
- Environmental Requirements:
- Temperature: 15 to 32°C
- Humidity: 5 to 90% noncondensing
- Barometric Pressure: 523 to 800 mmHg (69.7 to 106.7 kPa)
Specifications
USB (three ports), RS232 port, bar-code scanner port and RJ10/100 Base T Ethernet port | |
∼60 sec. | |
McAfee embedded control, firewall, verified system connection and encrypted patient data transfer via USB | |
Rating: 150 VA, Voltage: 100/240 VAC, Freq: 50/60 Hz |
Heparinized Whole Blood | |
One point every 30 minutes to a maximum of 60 minutes; Full calibration every 8 hours | |
pH only: 35 μL; Full menu: 90 μL | |
TUV-listed to IEC/EN 61010, 61010-2-101, 61010-2-081, CB Test Certificate, certified to CSA and UL |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction