Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1/AKT2/AKT3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKT1/AKT2/AKT3.
Supplier: Abnova Corporation PAB26831
Description
Sequence: P-E-Yp-L-ASpecifications
AKT1/AKT2/AKT3 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
P31749 (Gene ID : 207);P31751 (Gene ID : 208);Q9Y243 (Gene ID : 1000) | |
AKT1 | |
A synthetic peptide corresponding to residues surrounding Y315/Y316/Y312 of human AKT1/AKT2/AKT3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of AKT1/AKT2/AKT3. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
Rabbit | |
Affinity chromatography | |
RUO | |
207/208/10000 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction