Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (R)-DRF053 dihydrochloride
Potent CK1 inhibitor; also inhibits cyclin-dependent kinases
$358.44 - $1534.68
Specifications
Purity | >98% |
---|---|
Product Type | (R)-DRF053 dihydrochloride |
Molecular Weight (g/mol) | 490.43 |
Inhibitors | (R)-DRF053 dihydrochloride |
Description
Potent casein kinase 1 (CK1) inhibitor (IC50 = 14 nM). Also an ATP-competitive cyclin-dependent kinase (cdk) inhibitor (IC50 values are 0.093 - 0.29, 0.21 and 0.82 μM for cdk2, cdk5 and cdk7, respectively). Shown to inhibit amyloid-β production in N2A-APP695 cells.Specifications
>98% | |
(R)-DRF053 dihydrochloride | |
490.43 | |
(R)-DRF053 dihydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title