Learn More
OraSure Technologies Q.E.D.™ Saliva Alcohol Test, A-150
SDP
The Q.E.D.™ Saliva Alcohol Test is the 'on-the-spot leader' in the rapid, accurate and quantitative determination of alcohol in saliva. With a high correlation to blood-alcohol levels, Q.E.D. has a testing range of 0-145 mg/dL and provides quantitative results in minutes.
$301.00 - $996.00
Specifications
Certifications/Compliance CLIA-waived and DOT-approved alcohol testing
Type Saliva Alcohol Test
For Use With (Application) On-Site, Low-Cost Alternative to Breath or Blood Alcohol Testing
Test Time <5 min.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-222-108
SDP
View Documents
OraSure Technologies
31150B
30 Tests
Pack of 30 for $301.00
Add to Cart
23-222-109
SDP
View Documents
OraSure Technologies
31150C
100 Tests
Pack of 100 for $996.00
Add to Cart
Description
Description
  • Reliable, Fast, Simple
  • Non-invasive
  • Quantitative results in minutes
  • Reads like a thermometer
  • No instrumentation
  • Long shelf life
  • Built-in QA Spot™ assures quality control for every test
Specifications
Specifications
CLIA-waived and DOT-approved alcohol testing
Saliva Alcohol Test
On-Site, Low-Cost Alternative to Breath or Blood Alcohol Testing
<5 min.