Learn More
Perkin Elmer US LLC PTFE Tubing - 1 ft x 0.125 in O.D.
Supplier: Perkin Elmer US LLC 02506483
This tubing can be used to transport liquids, gasses, or air. It is compatible with TMA 4000, TGA PYRIS1/4000/8000 analyzers, as well as Clarus 400/500/600/690 instruments.