Learn More
Electron Microscopy Sciences ProScope HR2 QC Inspector
Supplier:  Electron Microscopy Sciences 68350642
Encompass_Preferred
Kit includes: HR base, 0-10x Lens, 50x Lens, HR round base stand, Proscope HR software, Screen Measurement software, Deluxe case.
Catalog No. 50-336-69