Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pro-Lab Diagnostics™ Microbank™ Freezer Storage Boxes
Suitable for collection and organization in low temperature freezers.
$1323.00
Catalog Number | Mfr. No. | Closure Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Color | Price | Quantity & Availability | |||||
22-286-222
|
Pro-Lab Diagnostics™
PL.169/B |
Blue |
Case of 24 for $1,323.00
|
|
|||||
22-286-223
|
Pro-Lab Diagnostics™
PL.169/R |
Red |
Case of 24 for $1,323.00
|
|
|||||
Description
- Manufactured from durable plastic
- Each freezer box holds up to 81 Microbank vials
- With “see through” lid, number locator printed screen, and tube collection device
- Available in blue or red
- Dimensions: 133 × 133 × 55mm
Specifications
Each | |
13.3 cm | |
5.23 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title