Learn More
Euthanex Preset CO2 Regulator (0-50 PSI) with 9' Hose & Quick-Disconnect
Supplier:  Euthanex EP1150P
Encompass
EP-1150-P Preset RegulatorPreset CO2 Regulator, 0 " 50 PSI Output. Connects directly to the CO2 cylinder with CGA-320 fitting. For use with equipment that requires a specific incoming pressure. Preset at factory. Includes a 9 foot hose with quick disconnect fitting.
Catalog No. NC2259360