Learn More
Thermo Scientific™ Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on Precision circulating water baths. Featuring advanced temperature controller.
$4049.00 - $5676.00
Specifications
Controller Type | Digital |
---|---|
Certifications/Compliance | UL, CE |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Includes | 2 Stainless Steel Gable Covers, 2 Diffuser Trays, and Rubber Duck |
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Electrical Requirements | Model | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Electrical Requirements | Model | Price | Quantity & Availability | ||
S37372
|
Thermo Scientific™
TSCIR19/EMD |
19L Circulating Water Bath | 19 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 19 |
Each for $4,049.00
|
|
||
S37373
|
Thermo Scientific™
TSCIR35/EMD |
35L Circulating Water Bath | 35 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 35 |
Each for $4,453.00
|
|
||
S37374
|
Thermo Scientific™
TSCIR89/EMD |
89L Circulating Water Bath | 89 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 89 |
Each for $5,676.00
|
|
||
Description
Thermo Scientific™ Precision™ circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
- Achieve enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Optimize scheduling with auto-on and auto-off timers
- Accommodate taller labware with new hinged lid and extended height
- Help prevent bath overheating and damage with low-fluid protection
- Easily operate and monitor with icon-based graphical display
- Protect your work with audible alarms
Specifications
Digital | |
Ambient +5° to 100°C | |
2 Stainless Steel Gable Covers, 2 Diffuser Trays, and Rubber Duck | |
4 | |
±0.05°C |
UL, CE | |
Monochrome LCD | |
304 Stainless Steel | |
±0.1°C | |
Global Voltage 100/115 V 200/230 V 50/60 Hz |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.