Learn More
Electron Microscopy Sciences Microscope Slide Polystyrene
Supplier:  Electron Microscopy Sciences 71889
Encompass_Preferred
Polystyrene microscope slides are 1" x 3". Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Catalog No. 50-949-666