Learn More
Polysciences, Inc. Polybead Microspheres 0.05 um, 10 mL
Supplier: Polysciences, Inc. 0869110
Polybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90um. Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.