Learn More
Waters Corp Polybutadiene Standards Kit
Supplier: Waters Corp WAT035709
A unique collection of 10 GPC standards (Gel Permeation Chromatography standards) for non-aqueous molecular weight determination. Polybutadiene Standards Kit
10 Vials, 0.5g in each vial containing the following moleculare weight standards:
1000, 3000, 7000, 10000, 30000, 70000, 100000, 300000, 700000, 1000000