Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Phosphorus Reagents for Hitachi™ Analyzers

Quantify inorganic phosphorus easily with system specific reagents

$481.00

Specifications

Certifications/Compliance CE Marked
Formulation Ammonium Molybdate >0.48mM, Sulfuric Acid <220mM with Surfactant
For Use With (Application) Quantitative determination of inorganic Phosphorus in Serum
Form Liquid
View More Specs

Includes: Reagent 1 and Reagent 2 when combined: Ammonium molybdate >0.48mM; Sulfuric acid <220mM; surfactant

Requires: Calibrator/standard


Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-666-379
View Documents
Pointe Scientific
HP9161200
8 x 100mL R1, 4 x 100mL R2
Pack of 12 for $481.00
Add to Cart
 
23-666-380
View Documents
Pointe Scientific
HP416R1
5 x 600 R1mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
23-666-377
View Documents
Pointe Scientific
HP716600
8 x 50mL R1, 4 x 50mL R2 Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
23-666-381
View Documents
Pointe Scientific
HP416R2
5 x 300 R2mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

  • Method: Phosphomolybdate, Endpoint
  • Format: Liquid (ready to use)
  • Linearity: 12mg/dL
  • Expected Values: 2.5 to 4.8mg/dL

Applications:

For the quantitative determination of inorganic phosphorus in serum on Hitachi analyzers

Specifications

Specifications

CE Marked
Ammonium Molybdate >0.48mM, Sulfuric Acid <220mM with Surfactant
Quantitative determination of inorganic Phosphorus in Serum
Liquid
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit