Learn More
Carolina Science & Math PINS MINUTEN 0.15MM DIA 500/P
SDP
Supplier:  Carolina Science & Math 654366
Encompass
Stainless steel. A very small, stiff pin ideal for use in humid conditions or with small marine specimens. 0.15 mm diameter, pack of 500.
Catalog No. NC9287493