Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Photo Numbers and Flags
SCI_ED

For 1–9 as numbers or flags.

Supplier:  Lynn Peavey Company 5816

Catalog No. S69030A


Add to Cart

Description

Description

Big, bold, and easy to read, these adhesive-backed numbers and flags will adhere to virtually any surface

Photo Numbers

  • Dimensions: 2 × 3 in.
  • Pack of five sets
Photo flags
  • Dimensions: 4 × 5 in.
  • Individual sets of nine

Specifications

Specifications

Photo Number
2 x 3 in.
Adhesive-backed
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit