Learn More
Regis Technologies Inc GC Regeant, Regis, PFPA (10 x 1 GM)
SDP
Supplier:  Regis Technologies Inc 1640110200
Encompass_Preferred
PFPA: Pentafluoropropionic Anhydride is an Acylation Reagent. PFPA is most commonly used for ECD and reacts with alcohols, amines, and phenols. Bases such as triethylamine and trimethylamine can be added to promote reactivity. PFPA is frequently used for the confirmation of drugs of abuse. PFPA derivatives require the lowest analysis temperatures.
Catalog No. 50-442-941
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.