Learn More
Biotang Inc PEG-LIOAC SOLUTION 250 ML, BP-257. 10 mM Tris-HCl (pH 8.0), 1 mM EDTA, 100 mM Lithium acetate, and 40% PEG 3350. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Supplier: Biotang Inc BTBB656
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PEG-LIOAC SOLUTION 250 ML, BP-257. 10 mM Tris-HCl (pH 8.0), 1 mM EDTA, 100 mM Lithium acetate, and 40% PEG 3350. Prepared in 18.2 megohms water and filtered through 1 micron filter.. This item has a minimum order quantity of 5 per supplier requirements for all the buffers.