Learn More
Paulson A-TAC™ Firefighter Goggles
Click to view available options
Frame color: Black
Description
- Polycarbonate, hard-coated/anti-fog lens
- Fit comfortably over prescription, sun, and safety glasses
- Silicon frame, black
- Indirect ventilation ports
- Can be placed on the helmet or worn on the head
Compliance
Meets ANSI Z87.1 standard for impact, penetration and chemical splash exposure
Specifications
Product Type | Goggle w/black frame and elastic strap |
Frame Color | Black |