Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutec Group Paddle Lab Blender - Masticator Compact
SDP

Used for solid and semisolid samples by applying pressure to samples within sterile, disposable bags

Supplier:  Neutec Group 900087

Request Bulk Order

Catalog No. 14-910-430


Add to Cart

Description

Description

  • Reduce work load on lab technician
  • Long tradition of reliability and robustness
  • Use metal springs to move paddles and not belt
  • Choice of digital or analog timer, compact size
  • Small bench space required due to its long rather than wide design
  • Viewing window option (panoramic models) for better monitoring of sample condition
  • Works with large selection of lab blending bags
  • Stroke: 10

Warranty and Services

3 years

Specifications

Specifications

3100 rpm
30 x 15 x 15 cm (11.8 x 5.9 x 5.9 in.)
Compact (80mL bag), Analog timer
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit