Learn More
Owen Mumford™ Autolet™ II Clinisafe and Autolet Lite Clinisafe Automatic Lancet Devices
SDP
Features easy ejection of both lancet and platform after each sample
Supplier:  Owen Mumford™ AT0900
Catalog No. 22-023357
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Two lancet devices provide a clean, rapid incision to a controlled depth, minimizing bruising and scarring of tissue.
Autolet™ Lite Clinisafe
  • Spring-loaded device conceals lancet inside the device before and after incision
  • After each patient sample, the lancet and platform (both potentially contaminated components) are ejected at the touch of a button
  • Easily operated with one hand, permitting patients to draw their own blood samples
  • Comes with 50 Unilet Superlite lancets, and instructions for use
Autolet II Clinisafe
  • Lancet fits into a spring-loaded arm and, when released by depressing a button, travels in a rapid arc to pierce the finger
  • Supplied with vinyl wallet, 10 Unilet Superlite lancets, 10 yellow platforms, and instructions
Accessories
  • Sterile, disposable Unilet Superlite lancets with round, ribbed for use with both Autolet devices
Product Suggestions
Product Suggestions