Learn More
PIP™ OTG Rimless Safety Glasses
Click to view available options
Clear lens
Description
- Over-the-glass design helps keep prescription eyewear users in compliance
- Can also be worn by itself as a regular safety glass
- Rimless frame design ensures an unobstructed view
- Adjustable length temples are rubber co-molded, offering the user a secure custom fit
- Ratchet temples for better angle adjustment
- Molded nose bridge design to fit the vast majority of users
- U-Fit™ spatula temples
- Meets ANSI Z87.1+
Specifications
Lens Tint | Clear |
Frame Color | Black |
Temple Style | Adjustable Length Ratchet Temples |
Temple Color | Black, Gray |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate / TPR |
UV Protection | 0.9999 |
Warning - Prop 65 - Reproductive Harm