Learn More
Electron Microscopy Sciences EMS Osteotome U.S. Army Premium, 6 mm, 6½" OAL
Supplier: Electron Microscopy Sciences 7799012
These U.S. Army pattern osteotomes, also called bone chisels, are used to score, cut, scrape, clean and sculpt the bone.
Size: 6 mm
OAL: 6½"