Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: B12
Supplier: Ortho-Clinical Diagnostics™ 1540525
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum and Plasma (EDTA or Heparin) | |
2mL VITROS™ Vitamin B12 calibrators 1 and 2 (Vitamin B12 in Buffer with Human Serum Albumin and antimicrobial agent), lot calibration card (for use with reagent pack 1, colored green), ancillary lot card (for use with reagent pack 2, col | |
2 |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Vitamin B12 Calibrator | |
Quantitative measurement of Vitamin B12 | |
1 Set/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction