Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OraSure Technologies OraQuick™ HCV Antibody Test
![SDP](/content/dam/fishersci/glyphs/sdp.png)
HCV Testing anytime, anywhere made easy. The OraQuick™ HCV test is the only point-of-care Hepatitis C test on the market today, detecting HCV antibodies in fingerstick and venipuncture whole blood. Yielding accurate results in as little as 20 minutes.
$575.00 - $2300.00
Specifications
Content And Storage | 2°C to 30°C |
---|---|
CE Marker | Yes |
Detectable Analytes | HCV |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-222-105
![]() |
OraSure Technologies
10010181 |
25 Tests |
Pack of 25 for $575.00
|
|
|||||
23-222-104
![]() |
OraSure Technologies
10010180 |
100 Tests |
Pack of 100 for $2,300.00
|
|
|||||
Description
- The OraQuick™ HCV test is FDA approved for detecting HCV antibodies in fingerstick and venipuncture whole blood.
- Our simple platform enables healthcare providers to deliver an accurate result in 20 minutes.
- The OraQuick™ HCV Rapid Antibody Test is comprised of both a single-use test device and vial containing a pre-measured amount of a buffered developer solution.
- The test consists of a sealed pouch with two separate compartments for each component.
- The developer solution facilitates the capillary flow of the specimen into the device and onto the assay strip.
- 25 Test Kit includes: 25 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
- 100 Test Kit includes: 100 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
Specifications
2°C to 30°C | |
Yes | |
HCV |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title