Learn More
Chemglass Life Sciences 70-Position Block for 10 x 75mm Round Bottom Tubes
SDP
Supplier:  Chemglass Life Sciences OP660030
Encompass_Preferred
70-Position Block for 10 x 75mm Round Bottom Tubes. Vial and tube holders for use in centrifugal vacuum evaporators. Blocks are designed to not only maximize evaporation but also to minimize the weight of the block, thereby increasing sample volumes. Constructed of aluminum, each block is black hard coated for resistance to organic solvents and laser etched with the vial/tube size.
Catalog No. 50-153-2707