Learn More
Diversified Spaces™ Oak Table with Plain Apron and Solid Epoxy Top: Leg Brace and Stretcher Option
Solid Epoxy tops are impervious to normal laboratory chemicals and heat. Diversified Spaces™ Oak Tables are ideal for high school and above with gas and other heat producing processes.
$1082.00 - $2432.00
Specifications
Adjustable | No |
---|---|
Certifications/Compliance | SEFA compliant and MAS Certified Green™ |
Finish | Chemical resistant, earth-friendly UV |
For Use With (Application) | High school and above with gas and other heat producing processes |
Includes | Leg Brace and Stretcher |
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
S41946
|
Diversified Spaces™
P7306K30S |
24 x 72 in. (60.96 x 182.88 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,507.00
|
|
|||
S41837
|
Diversified Spaces™
P7246K36S |
36 x 72 in. (91.44 x 182.88 cm) | 36 in. | Leg Brace and Stretcher |
Each for $2,219.00
|
|
|||
S41835
|
Diversified Spaces™
P7246K34S |
36 x 72 in. (91.44 x 182.88 cm) | 34 in. | Leg Brace and Stretcher |
Each for $2,219.00
|
|
|||
S42009
|
Diversified Spaces™
P7406K30S |
42 x 72 in. (106.68 x 182.88 cm) | 30 in. | Leg Brace and Stretcher |
Each for $2,359.00
|
|
|||
S41956
|
Diversified Spaces™
P7306K36S |
24 x 72 in. (60.96 x 182.88 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,580.00
|
|
|||
S41953
|
Diversified Spaces™
P7306K34S |
24 x 72 in. (60.96 x 182.88 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,580.00
|
|
|||
S42013
|
Diversified Spaces™
P7406K34S |
42 x 72 in. (106.68 x 182.88 cm) | 34 in. | Leg Brace and Stretcher |
Each for $2,432.00
|
|
|||
S42239
|
Diversified Spaces™
P7806K30S |
42 x 54 in. (106.68 x 137.16 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,925.00
|
|
|||
S42167
|
Diversified Spaces™
P7606K36S |
24 x 60 in. (60.96 x 152.4 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,396.00
|
|
|||
S42297
|
Diversified Spaces™
P7906K30S |
42 x 60 in. | 30 in. | Leg Brace and Stretcher |
Each for $2,093.00
|
|
|||
S42243
|
Diversified Spaces™
P7806K34S |
42 x 54 in. (106.68 x 137.16 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,999.00
|
|
|||
S42153
|
Diversified Spaces™
P7606K30S |
24 x 60 in. (60.96 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,322.00
|
|
|||
S42015
|
Diversified Spaces™
P7406K36S |
42 x 72 in. (106.68 x 182.88 cm) | 36 in. | Leg Brace and Stretcher |
Each for $2,432.00
|
|
|||
S42247
|
Diversified Spaces™
P7806K36S |
42 x 54 in. (106.68 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,999.00
|
|
|||
S42305
|
Diversified Spaces™
P7906K36S |
42 x 60 in. | 36 in. | Leg Brace and Stretcher |
Each for $2,169.00
|
|
|||
S42617
|
Diversified Spaces™
P7906K34S |
42 x 60 in. | 34 in. | Leg Brace and Stretcher |
Each for $2,169.00
|
|
|||
S42728
|
Diversified Spaces™
P7136K34S |
30 x 54 in. (76.2 x 137.16 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,565.00
|
|
|||
S42723
|
Diversified Spaces™
P7126K34S |
30 x 48 in. (76.2 x 121.92 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,392.00
|
|
|||
S42764
|
Diversified Spaces™
P7146K34S |
30 x 60 in. (76.2 x 152.4 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,627.00
|
|
|||
S47489
|
Diversified Spaces™
P7106K30S |
24 x 48 in. (60.96 x 121.92 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,176.00
|
|
|||
S47495
|
Diversified Spaces™
P7106K34S |
24 x 48 in. (60.96 x 121.92 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,250.00
|
|
|||
S47500
|
Diversified Spaces™
P7106K36S |
24 x 48 in. (60.96 x 121.92 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,250.00
|
|
|||
S47626
|
Diversified Spaces™
P7126K30S |
30 x 48 in. (76.2 x 121.92 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,316.00
|
|
|||
S47636
|
Diversified Spaces™
P7126K36S |
30 x 48 in. (76.2 x 121.92 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,392.00
|
|
|||
S47571
|
Diversified Spaces™
P7116K36S |
42 x 48 in. (106.68 x 121.92 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,822.00
|
|
|||
S47705
|
Diversified Spaces™
P7136K30S |
30 x 54 in. (76.2 x 137.16 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,492.00
|
|
|||
S47719
|
Diversified Spaces™
P7136K36S |
30 x 54 in. (76.2 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,565.00
|
|
|||
S47853
|
Diversified Spaces™
P7146K30S |
30 x 60 in. (76.2 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,553.00
|
|
|||
S47867
|
Diversified Spaces™
P7146K36S |
30 x 60 in. (76.2 x 152.4 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,627.00
|
|
|||
S47995
|
Diversified Spaces™
P7156K30S |
30 x 72 in. | 30 in. | Leg Brace and Stretcher |
Each for $1,684.00
|
|
|||
S48003
|
Diversified Spaces™
P7156K36S |
30 x 72 in. | 36 in. | Leg Brace and Stretcher |
Each for $1,757.00
|
|
|||
S48001
|
Diversified Spaces™
P7156K34S |
30 x 72 in. | 34 in. | Leg Brace and Stretcher |
Each for $1,757.00
|
|
|||
S48087
|
Diversified Spaces™
P7166K30S |
21 x 28 in. (53.34 x 121.92 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,082.00
|
|
|||
S48175
|
Diversified Spaces™
P7176K34S |
36 x 48 in. (91.44 x 121.92 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,664.00
|
|
|||
S48180
|
Diversified Spaces™
P7176K36S |
36 x 48 in. (91.44 x 121.92 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,664.00
|
|
|||
S48258
|
Diversified Spaces™
P7186K34S |
21 x 54 in. (53.34 x 137.16 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,249.00
|
|
|||
S48338
|
Diversified Spaces™
P7196K34S |
36 x 54 in. (91.44 x 137.16 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,849.00
|
|
|||
S48265
|
Diversified Spaces™
P7186K36S |
21 x 54 in. (53.34 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,249.00
|
|
|||
S48334
|
Diversified Spaces™
P7196K30S |
36 x 54 in. (91.44 x 137.16 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,776.00
|
|
|||
S48475
|
Diversified Spaces™
P7206K30S |
24 x 54 in. (60.96 x 137.16 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,209.00
|
|
|||
S48342
|
Diversified Spaces™
P7196K36S |
36 x 54 in. (91.44 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,849.00
|
|
|||
S48489
|
Diversified Spaces™
P7206K36S |
24 x 54 in. (60.96 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,283.00
|
|
|||
S48581
|
Diversified Spaces™
P7216K30S |
21 x 60 in. (53.34 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,272.00
|
|
|||
S48588
|
Diversified Spaces™
P7216K34S |
21 x 60 in. (53.34 x 152.4 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,347.00
|
|
|||
S41831
|
Diversified Spaces™
P7246K30S |
36 x 72 in. (91.44 x 182.88 cm) | 30 in. | Leg Brace and Stretcher |
Each for $2,144.00
|
|
|||
S48664
|
Diversified Spaces™
P7226K30S |
36 x 60 in. (91.44 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,871.00
|
|
|||
S48595
|
Diversified Spaces™
P7216K36S |
21 x 60 in. (53.34 x 152.4 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,347.00
|
|
|||
S48730
|
Diversified Spaces™
P7236K30S |
21 x 72 in. (53.34 x 182.88 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,462.00
|
|
|||
S48672
|
Diversified Spaces™
P7226K36S |
36 x 60 in. (91.44 x 152.4 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,947.00
|
|
|||
S48737
|
Diversified Spaces™
P7236K34S |
21 x 72 in. (53.34 x 182.88 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,536.00
|
|
|||
S48740
|
Diversified Spaces™
P7236K36S |
21 x 72 in. (53.34 x 182.88 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,536.00
|
|
|||
Description
- 1 in. Epoxy Table Surface
- 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
- Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Includes leg braces and stretcher
- Limited Lifetime Warranty
- Made in the USA
- Minimal assembly required; legs need to be attached
- Solid Oak Construction
Specifications
No | |
Chemical resistant, earth-friendly UV | |
Leg Brace and Stretcher | |
226.79 kg | |
Solid Epoxy | |
Red Oak |
SEFA compliant and MAS Certified Green™ | |
High school and above with gas and other heat producing processes | |
500 lb. | |
Plain Apron | |
Plain Apron Table |