Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ pH 3-7 IEF Protein Gels, 1.0 mm

Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI).

$248.00 - $249.00

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 5%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type IEF
View More Specs

Products 2
Catalog Number Mfr. No. Wells Price Quantity & Availability  
Catalog Number Mfr. No. Wells Price Quantity & Availability  
50-591-656
Encompass_Preferred
View Documents Promotion Details
Invitrogen™
EC6645BOX
10-well
Each for $249.00
Add to Cart
 
EC66452BOX
Encompass_Preferred
View Documents Promotion Details
Invitrogen™
EC66452BOX
12-well
Each for $248.00
Add to Cart
 
Description

Description

Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI). The pI is the pH at which a protein has no net charge and does not move in an electric field. Vertical IEF gels effectively create a pH gradient so proteins separate according to their unique pI.

IEG gels can be used to readily detect minor changes in a protein due to deamination, phosphorylation, or glycosylation, and can resolve different proteins of similar size that cannot be resolved on standard SDS-PAGE gels. Pre-focusing is not required. Total run time is approximately 2.5 hours.

Formulation
Novex™ IEF gels are 5% polyacrylamide and consist of high-purity acrylamide, bisacrylamide, TEMED, APS, ultrapure water, and 2% ampholytes. They do not contain denaturing reagents.

Recommended buffers
IEF is a sensitive technique which is affected by many factors. Our optimized, premixed IEF buffers reduce variability and help you obtain consistent results.

Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
IEF
Isoelectric Focusing
Novex™
Wet Ice
8 cm
5%
1.0 mm
3.5 to 6.8
pI (Charge)
5 Gels/Box
Store at 2°C to 8°C. Do not freeze.
Safety and Handling

Safety and Handling

ShelfLife : 6 Months

SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit