Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ BL21 Competent Cells
BL21 host strain is the most widely used host background and has the advantage of being deficient in both lon and ompT proteases.
Specifications
Contains F' Episome | Lacks F' Episome |
---|---|
Efficiency | Guaranteed efficiency: >2 x 107 cfu/μg |
Content And Storage | ≤-70°C |
Blue/White Screening | No |
Description
BL21 is the most widely used host background and has the advantage of being deficient in bothSpecifications
Lacks F' Episome | |
Guaranteed efficiency: >2 x 107 cfu/μg | |
≤-70°C | |
No |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title