Learn More
Immunology Consultants Normal Dog Serum, 100 ml
Supplier:  Immunology Consultants RS40100ML
Encompass_Preferred
Normal dog serum was collected from healthy dogs and sterile filtered. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2175