Learn More
Mopec Non-Elevating Autopsy Table with Integral Right Wing
SDP
Includes all of the features necessary to perform an autopsy along with an added integrally constructed wing. Mopec™ Non-Elevating Autopsy Table with Integral Right Wing is an L-shaped table with pedestal design that includes standard downdraft ventilation system.
Supplier:  Mopec CE600
Request Bulk Order
Catalog No. 22-444-030
Add to Cart
Description
Description
  • All stainless steel construction
  • 14 gauge stainless steel table top, sink and grid plates
  • 18 gauge stainless steel pedestal
  • All type 304 stainless steel with #4 finish
  • Large utility pedestal with smaller storage support pedestal and support section
  • Hydro aspirator designed with reversing flow valve
  • With hose hanger and 8 foot of clear PVC tubing
  • Cold water control valve with vacuum brake swing spout
  • 4 interchangeable removable grid plates, stainless steel construction with 0.5 in. recess and 0.375 in. diameter holes on 2 in. square centers
  • Heavy duty brass chrome plated hand piece
  • Removable pedestals if need for easier installation
Warranty and Services
1 Year Warranty
Specifications
Specifications
Stationary Pedestal Table
Down Draft
10 in.
25.4 cm
100 in.
37 in.
93.98 cm
254 cm
24 in.
76.2 cm
50.8 cm
2 GFCI Duplex Outlet with Waterproof Cover
90 in.
228.6 cm
37 in.
30 in.
93.98 cm
12 in.
30.48 cm
20 in.
60.96 cm
Product Suggestions
Product Suggestions