Learn More
Organomation 24 position N-EVAP
SDP
Supplier:  Organomation 11250NT
Encompass_Preferred
Request Bulk Order
The 24 position N-EVAP (Cat# 11250) comes with an option code (-NT) that makes it PFAS free. The 24 position N-EVAP offers the most customizable options for your sample preparation needs. The standard sample holder can accommodate samples with an outside diameter of 10-30 mm. However, with the optional 8 position large sample holder, vials with an outside diameter of 30-70 mm can be processed. The 24 position N-EVAP test tube evaporator can also be upgraded to include digital controls for heat and time with the addition of the side control box (Option code -CB). These economical units provide controlled concentration of sample vials through the application of nitrogen gas within a heated water bath. The dual band spring hoist assembly provides a simple and effective means of removing samples from the heated dry bath.
Catalog No. NC2301756