Learn More
Enzo Life Sciences Netrin-1 (mouse), (recombinant) (10µg)
Supplier: Enzo Life Sciences ALX5221242010
Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse netrin-1 (aa 27-604) is fused at the C-terminus to a FLAG-tag. Specificity: Binds to human and mouse UNC5B. Long Term Storage: -20°C. Applications: ELISA .