Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Multifibren™ U Reagent
Quantitative determination of fibrinogen in plasma.
$505.00 - $1187.00
Specifications
Type | Hemostasis Reagent |
---|---|
For Use With (Application) | Fibrinogen Testing |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-044-709
|
Siemens Healthineers
10446689 |
10 x 2 mL |
Pack of 10 for $505.00
|
|
|||||
23-044-710
|
Siemens Healthineers
10446691 |
10 x 5 mL |
Pack of 10 for $1,187.00
|
|
|||||
Description
Multifibren™ U Reagent is a bovine thrombin reagent used in the modified Clauss determination of fibrinogen for the detection of hereditary or acquired hypo- and hyperfibrinogenemia and dysfibrinogenemia. The reagent is insensitive to heparin up to 2.0 U/mL and has a wide measuring range.Specifications
Hemostasis Reagent | |
Fibrinogen Testing |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title