Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mouse IgG2b Isotype Control, PE, Invitrogen™
Mouse Isotype Control
Supplier: Invitrogen™ MG2B04
This item is not returnable.
View return policy
Description
Mouse IgG2b Isotype Control for Flow
Mouse IgG2b Control antibody specifically detects Mouse IgG2b in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG2b | |
PE | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG2b | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction