Learn More
Ted Pella Inc MOLD PELCO FLAT EMBEDDING
Supplier: Ted Pella Inc 1052
PELCO Flat Embedding Mold, Mold Size: 105 x 70 x 7mm thick, Special Cavities: 1 trough - 1cm x 1.7cm x 3mm deep, 2 troughs - 1cm x 3cm x 3mm deep, Numbering: A1- A6, B1-B6, C1-C6, D1-D6, Number of Cavities: 24, anumeric self-impression cavi