Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSecure™ MobileAid™ Easy-Roll Modular Trauma First Aid Station
SCI_ED

Extensive 260-piece mobile Modular Trauma First Aid Station with 4 additional Bleeding Control Kits to treat bleeding wounds from accidents, disasters, or active shooter situations. 

$1159.00 - $1399.00

Specifications

Product Type Trauma First Aid Kit
For Use With (Application) First Aid/Bleed Control
Length (Metric) 46.99 cm
Certifications/Compliance ISEA Z308.1-2015 Class A, Type II
Color Red
View More Specs

Products 3
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
Catalog Number Mfr. No. Model Includes Price Quantity & Availability  
S29686
SCI_ED
View Documents
LifeSecure™
31520
MobileAid BleedStop Immediate-Response 100 Bleeding Control Kits 4 Immediate-Response 100 Bleeding Control Kits, SWAT-T tourniquets, BloodStopper compress bandages, QuickClot blood clotting gauze, Petroleum gauze for chest seals, and other bleeding wound supplies
Each for $1,159.00
Add to Cart
 
S29687
SCI_ED
View Documents
LifeSecure™
31525
MobileAid BleedStop Immediate-Response 200 Bleeding Control Kits 4 Immediate-Response 200 Bleeding Control Kits, SOFT-W tourniquets, BloodStopper compress bandages, QuickClot blood clotting gauze, Petroleum gauze for chest seals, and other bleeding wound supplies
Each for $1,280.00
Add to Cart
 
S29688
SCI_ED
View Documents
LifeSecure™
31530
MobileAid BleedStop Immediate-Response 300 Bleeding Control Kits 4 Immediate-Response 300 Bleeding Control Kits, SOFT-W tourniquets, BloodStopper compress bandages, QuickClot blood clotting gauze, Chest seals, and other bleeding wound supplies
Each for $1,399.00
Add to Cart
 
Description

Description

  • Compact dual-wheel mobile station
  • Organized in quick-response first aid modules for rapid response and treatment
  • Includes a flexible evacuation stretcher and First Aid Flag
  • Integrated seat and gear platform allow patient to sit and be treated or can be used to haul additional equipment (e.g. AED)
Specifications

Specifications

Trauma First Aid Kit
46.99 cm
Red
Up to 50 People
48.26 cm
First Aid/Bleed Control
ISEA Z308.1-2015 Class A, Type II
1 Ea.
18.5 x 19.5 x 14 in.
35.56 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit