Learn More
Fisherbrand™ Mini Centrifuge PROMO
Tool-free rotors and quick-spin options make this Mini Centrifuge fast and easy to use. The locking lid is functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction.
Catalog No. 12-006-901S24
Add to Cart
Description
Description
  • 5 Year Warranty
  • A locking lid for added protection when the unit is in use
  • A larger lid to accommodate longer and skirted tubes, as well as taller tube lids
  • A braking function to quickly stop the unit when powered off or when the lid is opened
  • Meets the latest applicable North American and EU safety requirements
  • Weighted rubberized base eliminates potential movement and further dampens vibration and noise
Specifications
Specifications
6 Tubes
2,000 x g
5.03 in.
Mini Centrifuge
Yes
50/60 Hz
6,000 rpm
No
128 mm
Two Rotors, Tube Rack, Storage Case, O-rings, Tube Adapters, Low Voltage, Double Insulated Power Adapter with Four Interchangeable Plugs
104 x 128 x 153 mm
100/240 V
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories