Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millex™-HV Sterile Syringe Filter Unit, PVDF, 0.45 μm (Research Use Only)

Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0 μm pore sizes.

$275.00 - $935.00

Specifications

Autoclavable Not Autoclavable
Color Yellow
Disposable Single-use
Material Durapore PVDF
Pore Size 0.45 μm
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Quantity Tip Material Pressure Rating Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Quantity Tip Material Pressure Rating Price Quantity & Availability  
SLHVR33RS
View Documents Promotion Details
MilliporeSigma™
SLHVR33RS
3.3 cm 50Pack Modified Acrylic 145 psi
Pack of 50 for $275.00
Add to Cart
 
SLHVR13SL
View Documents Promotion Details
MilliporeSigma™
SLHVR13SL
1.3 cm 100Pack Polyethylene 100 psi
Pack of 100 for $482.00
Edge
Add to Cart
 
SLHVR33RB
View Documents Promotion Details
MilliporeSigma™
SLHVR33RB
3.3 cm 250Pack Modified Acrylic 145 psi
Pack of 250 for $935.00
Add to Cart
 
Description

Description

Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0 μm pore sizes.

Sterile Millex filter units are available in a larger 33 mm housing with either MCE, PVDF or PES membranes.

Faster flow rate:
The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.

Higher operating pressure:

Millex filters have a maximum housing pressure of 10.5 bar (150 psig), which means you can filter solutions faster than before.

Color-coded:

The color-coded band on the Millex housing clearly indicates which membrane is inside.

Manufactured for reliable performance:

Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filter during assembly. A Certificate of Quality included in each box describes Millipore's quality standards in detail.

Choice of membranes:
Millex filters are available with three membranes: Millipore Express™ (PES) membrane, mixed cellulose esters (MCE), and Durapore (PVDF).

  • Millex-GP filters feature Millipore Express (PES) membrane, which provides a unique combination of speed and six times less protein binding than competitive PES membranes. Millex-GP filters are recommended for sterile filtering protein solutions, tissue culture media, buffers and additives.
  • Millex filters with Durapore™ (PVDF) membrane are the lowest protein-binding syringe filters available. Use them to filter protein solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.
  • Millex-MF filters feature mixed cellulose esters (MCE) membrane, which provides reliable, general purpose filtration of water or other aqueous solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.
    Applications:
    Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components.
Specifications

Specifications

Not Autoclavable
Single-use
0.45 μm
Sterilization of tissue culture media, protein solutions or aqueous solutions;Low Protein Binding
Female Luer Lok
113°F
113°F
Syringe Filter
Yellow
Durapore PVDF
Sterile
PVDF
Male Luer Slip
45°C
45°C
100 mL
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit