Learn More
Microbiologics™ Bordetella parapertussis ATCC™ 15311™†
SDP
For culture purposes and QC applications
Specifications
Species Bordetella parapertussis
Product Type KWIK-STIK
View More Specs
Products 2
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
Catalog Number Mfr. No. ATCC® Product Type Price Quantity & Availability  
23-001-379
SDP
View Documents
Microbiologics™
0842P
15311 KWIK-STIK
Pack of 2 for $197.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-364
SDP
View Documents
Microbiologics™
0842K
ATCC 15311 KWIK-STIK
Pack of 6 for $438.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Bordetella parapertussis in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • ATCC™ 15311™†
Specifications
Specifications
Bordetella parapertussis
KWIK-STIK
Safety and Handling
Safety and Handling
ShelfLife : 6 to 24 Months