Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog Number M207525G
Click to view available options
Quantity:
5 g
25 g
2971-79-1
C7H13NO2
143.19
MFCD00190578
RZVWBASHHLFBJF-UHFFFAOYSA-N
methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
424914
methyl piperidine-4-carboxylate
COC(=O)C1CCNCC1
2971-79-1
C7H13NO2
143.19
MFCD00190578
RZVWBASHHLFBJF-UHFFFAOYSA-N
methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
424914
methyl piperidine-4-carboxylate
COC(=O)C1CCNCC1
Color Yellow
Boiling Point 93°C
Quantity 25 g
Formula Weight 143.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl 4-Piperidinecarboxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit