Learn More
Parker MERV 8 Pre-Filter
SDP
MERV 8 pleated glass fiber media pre-filter for trapping large particles before they enter the main filter. The pre-filter also helps to evenly distribute air across the main filter to maximize efficiency and extend filter life.
Supplier:  Parker DPHT101AGS
Catalog No. 14-950-431
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • MERV 8 pleated glass fiber media prefilter mounted inside a metal frame
  • Filter is approximately 1 in. thick
  • Traps large particles preventing entry into main filter
  • Evenly distributes air across the main filter to maximize efficiency and extend filter life
  • One filter is required every 3 months, or when the change filter light is illuminated
Specifications
Specifications
Metal
Pleated Glass Fiber
Pre-filter
MERV 8
Trapping large particles to prevent entry into the main filter
3 Months
Product Suggestions
Product Suggestions