Learn More
Mercedes Medical Glass Coverslips
Available in different shapes, thickness and sizes
Supplier: Mercedes Medical MER R2440
Specifications
Coverslip | |
Borosilicate Glass | |
40 mm | |
24 x 40 mm |
Rectangle | |
24 mm | |
#1 (0.13 to 0.16 mm) | |
Leica™ recommended |