Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
4X-Large
4X-Large (Long)
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- Extra comfortable fabric
- Underarm gusset for improved breathability
- Rib-knit crewneck collar
- Single spade chest pocket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
Size | X-Large (Long) |
Gender | Men's |
Material | FR Cotton |
Color | Navy |
Neck Style | Hemmed Crew |
Sleeve Style | Long Sleeve |
Disposable | Reusable |
No. of Pockets | 1 |
Hazard Risk Category | Category 2 |
Arc Flash Rating | 8.9 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction