Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt
Click to view available options
Size:
2X-Large
2X-Large (Long)
3X-Large
Large
Large (Long)
Medium
Small
X-Large
X-Large (Long)
Description
- Lightweight fabric flexes for added comfort and range of movement
- Contrasting pattern on sleeve for an athletic look
- Curved flatlock seams for optimal fit
- Unique front pocket design with pencil stall
- Ribbed collar
- CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric
Specifications
Size | Medium |
Gender | Men's |
Material | Cotton, Spandex |
Color | Navy |
Neck Style | Hemmed Crew |
Sleeve Style | Long Sleeve |
Construction | Knit |
Closure Type | Button |
Disposable | Reusable |
No. of Pockets | 1 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction