Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Meet ASTM™ Dart/Elmendorf and DOT CFR 49 173-197 requirements.
$30.50 - $281.00
Specifications
Product Type | Biohazardous Waste Bag |
---|---|
Certifications/Compliance | ASTM Dart |
Color | Red, Black |
Includes | Biohazardous or infectious waste with biohazard symbol |
Material | LLDPE |
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Thickness (English) | Price | Quantity & Availability | |||
14-828-192
|
Medegen
R108M |
12 to 16 gal. | 25 x 34 in. (63.5 x 86.36cm) | 1.20 mil |
Case of 300 for $209.00
|
|
|||
14-828-197
|
Medegen
R116 |
7 to 10 gal. | 23 x 23 in. (58.42 x 58.42cm) | 1.50 mil |
Case of 300 for $164.00
|
|
|||
14-828-201
|
Medegen
R172M |
44 gal. | 38 x 45 in. (96.52 x 114.3cm) | 1.20 mil |
Case of 200 for $281.00
|
|
|||
14-828-223
|
Medegen
RD630 |
2 to 3 gal. | 14.5 x 19 in. (36.83 x 48.26cm) | 1.25 mil |
Case of 200 for $52.75
|
|
|||
14-828-222
|
Medegen
RD610 |
1 to 3 gal. | 11 x 14.25 in. (27.94 x 36.195cm) | 1.25 mil |
Case of 200 for $30.50
|
|
|||
14-828-224
|
Medegen
RD650 |
5 to 6 gal. | 16 x 24 in. (40.64 x 60.96cm) | 1.25 mil |
Case of 200 for $54.00
|
|
|||
14-828-194
|
Medegen
R110 |
44 gal. | 38 x 45 in. (96.52 x 114.3cm) | 1.75 mil | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
14-828-199
|
Medegen
R117M |
30 gal. | 30.5 x 41 in. (77.47 x 104.14cm) | 1.20 mil |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||
14-828-198
|
Medegen
R117 |
30 gal. | 30.5 x 41 in. (77.47 x 104.14cm) | 1.50 mil | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- Red bag with black printing
- Biohazardous/Infectious Waste label with Biohazard symbol printed on bag
Specifications
Biohazardous Waste Bag | |
Red, Black | |
LLDPE |
ASTM Dart | |
Biohazardous or infectious waste with biohazard symbol |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title