Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Alcohol Prep-Pad Wipes

For disinfecting and preparation of skin prior to injections.

$4.85 - $9.28

Specifications

Product Type Alcohol Prep Pad
Composition isopropyl alcohol 70%
For Use With (Application) For disinfecting and preparation of skin prior to injections
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
19-090-834
View Documents
Medique
22133
100/Pk.
Each for $9.28
Case of 20 EA for $131.60
Edge
Add to Cart
 
19-090-833
View Documents
Medique
22150
50/Box
Each for $4.85
Case of 20 EA for $87.00
Edge
Add to Cart
 
Description

Description

  • Towelettes saturated with isopropyl alcohol 70%
Specifications

Specifications

Alcohol Prep Pad
isopropyl alcohol 70%
For disinfecting and preparation of skin prior to injections
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit