Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mechanix Wear™ Material4X™ Original™ Mechanics Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Seamless Material4X™ palms deliver resistance to tears and abrasion
- Form-fitting TrekDry™ material helps keep hands cool and comfortable
- Thermoplastic Rubber (TPR) closure with hook and loop provides a secure fit
- Reinforcement panels in high wear areas for additional durability
- Machine washable
Specifications
Back Material | TrekDry™ |
Color | Black, Gold |
Hand | Specific |
Material | Synthetic Leather |
Product Type | Mechanic's Gloves |
Size | 2X-Large |
For Use With (Application) | Construction, Maintenance Repairs, Equipment Operation, Woodwork, Landscaping, and DIY Home Improvement |
Glove Cuff Style | Hook and Loop |
Series | Original™ MG4X |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction