Learn More
Electron Microscopy Sciences Measuring Magnifier Mag6
Supplier: Electron Microscopy Sciences 68055
A hand-held magnifier for measurement and inspection. Their construction make them ideal aids for anyone concerned with quality control, inspection, drafting, photography, etc. The clear base allows ample surface light to fall on the subject being measured and the eye lens can be focused to suit individual sight. Interchangeable graticules fitted in the base ring permit a variety of measurements to be made quickly and accurately. Comes complete with a leather case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.