Learn More
MCR Safety Crews™ Glasses Standard Goggles
Click to view available options
Small
Description
- Made of PVC for comfortable fit over prescription eyewear
- Polycarbonate lens filters 99.9% of harmful UV rays
- Latex-free elastic strap
- Duramass™ scratch-resistant coating
Specifications
Product Type | Safety goggles |
Frame Color | Clear |
Size | Small |
Lens Material | Polycarbonate |
Frame Material | PVC |