Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Breakaway Safety Glasses Cord
Click to view available options
Black
Description
- Eyeglass Cord
- Black, slips over temple tips
- Stylish cord helps you keep up with your glasses
Specifications
Color | Black |
Product Type | Breakaway Cord |
For Use With (Application) | To keep glasses in reach when not in use |
Material | Nylon |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction